Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017934.5 | NP_060404.3 | 26 | 3337 | ||
XM_005248729.4 | XP_005248786.1 | 26 | 3334 | |||
XM_011535918.2 | XP_011534220.1 | 23 | 2993 | |||
XM_017010989.1 | XP_016866478.1 | 28 | 3479 | |||
XM_017010990.1 | XP_016866479.1 | 25 | 3077 | |||
GenBank mRNA | AJ303102.1 | - | 26 | 3124 | ||
AL161957.1 | - | 4 | 400 | |||
BC008909.2 | - | 1 | 65 | |||
BC081569.1 | - | 4 | 380 | |||
BC137488.1 | - | 26 | 3217 | 9021488 | ||
DQ924532.1 | - | 26 | 3111 |