Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083907.2 | NP_001077376.2 | 14, 15 | 2283 | ||
NM_001127507.2 | NP_001120979.2 | 13, 14 | 2115 | |||
NM_017935.4 | NP_060405.4 | 14, 15 | 2514 | |||
XM_017008337.1 | XP_016863826.1 | 14, 15 | 2261 | |||
GenBank mRNA | AB063170.1 | - | 13, 14 | 2150 | ||
AK000713.1 | - | 14, 15 | 2232 | |||
AK091523.1 | - | 18, 19 | 2615 | |||
AK093024.1 | - | 2, 3 | 1780 | |||
AK292133.1 | - | 14, 15 | 2358 | |||
AK301936.1 | - | 5, 6 | 793 | |||
AL834291.1 | - | 14, 15 | 2333 | |||
BC032241.1 | - | 14, 15 | 2382 | 5214147 | ||
EU051376.1 | - | 13, 14 | 1853 |