Search
Search

CHKA| Gene Symbol: CHKA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277.2 | NP_001268.2 | 5, 6 | 977 | ||
| NM_212469.1 | NP_997634.1 | 4, 5 | 923 | |||
| XM_017017147.1 | XP_016872636.1 | 4, 5 | 861 | |||
| XM_017017148.1 | XP_016872637.1 | 4, 5 | 2333 | |||
| GenBank mRNA | BC036471.2 | - | 4, 5 | 801 | 5265994 | |
| D10704.1 | - | 5, 6 | 787 | |||