Search
Search

CHKA| Gene Symbol: CHKA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277.2 | NP_001268.2 | 10, 11 | 1440 | ||
| NM_212469.1 | NP_997634.1 | 9, 10 | 1386 | |||
| XM_017017147.1 | XP_016872636.1 | 9, 10 | 1324 | |||
| XM_017017148.1 | XP_016872637.1 | 9, 10 | 2796 | |||
| GenBank mRNA | BC036471.2 | - | 9, 10 | 1264 | 5265994 | |
| D10704.1 | - | 10, 11 | 1250 | |||