Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CHN2 SIRNA Assays ›
Gene Symbol:
CHN2
Gene Name:
chimerin 2
Gene Aliases:
ARHGAP3, BCH, CHN2-3, RHOGAP3
Chromosome Location:
Chr.7: 29146547 - 29514335 on Build GRCh38
UniGene ID:
Hs.654611
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s3020
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CHN2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039936.2
NP_001035025.1
6, 7
1225
NM_001293069.1
NP_001279998.1
13, 14
1496
NM_001293070.1
NP_001279999.1
13, 14
1710
NM_001293071.1
NP_001280000.1
11, 12
1615
NM_001293072.1
NP_001280001.1
12, 13
1428
NM_001293073.1
NP_001280002.1
6, 7
1090
NM_001293075.1
NP_001280004.1
5, 6
952
NM_001293076.1
NP_001280005.1
5, 6
1051
NM_001293077.1
NP_001280006.1
4, 5
1009
NM_001293078.1
NP_001280007.1
4, 5
913
NM_001293080.1
NP_001280009.1
5, 6
1087
NM_001293081.1
NP_001280010.1
6, 7
740
NM_004067.3
NP_004058.1
12, 13
1671
NR_120525.1
4, 5
831
XM_011515105.2
XP_011513407.1
15, 16
1551
XM_011515106.2
XP_011513408.1
14, 15
1512
XM_011515107.2
XP_011513409.1
14, 15
2294
XM_017011721.1
XP_016867210.1
16, 17
1569
XM_017011722.1
XP_016867211.1
15, 16
2312
GenBank mRNA
AK026415.1
-
6, 7
1225
AK293959.1
-
13, 14
1290
AK294032.1
-
12, 13
1428
AK294216.1
-
6, 7
740
AK294271.1
-
13, 14
1497
AK301235.1
-
6, 7
749
AK313021.1
-
12, 13
1445
AK316030.1
-
6, 7
749
BC112155.1
-
12, 13
1416
8327623
EU732751.1
-
4, 5
432
EU732752.1
-
6, 7
691
EU732753.1
-
5, 6
553
EU732754.1
-
5, 6
652
EU732755.1
-
4, 5
514
EU732756.1
-
4, 5
610
EU732758.1
-
5, 6
688
EU732760.1
-
11, 12
1178
EU732762.1
-
9, 10
802
EU732763.1
-
5, 6
202
EU732764.1
-
6, 7
406
EU732765.1
-
7, 8
376
GQ924106.1
-
13, 14
1497
U28926.1
-
12, 13
1468
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of signal transduction
intracellular signal transduction
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
SH3/SH2 adaptor activity
GTPase activator activity
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-gzxth:80/10.239.166.29:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0