Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LARP1B SIRNA Assays ›
Gene Symbol:
LARP1B
Gene Name:
La ribonucleoprotein domain family member 1B
Gene Aliases:
LARP2
Chromosome Location:
Chr.4: 128060655 - 128222931 on Build GRCh38
UniGene ID:
Hs.657067
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s30245
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LARP1B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001278604.1
NP_001265533.1
5
464
NM_018078.3
NP_060548.2
5
464
NM_032239.3
NP_115615.2
5
464
NM_178043.2
NP_835144.1
5
464
XM_011532057.2
XP_011530359.2
5
1266
XM_011532058.2
XP_011530360.2
5
1265
XM_011532059.2
XP_011530361.2
5
1266
XM_011532060.2
XP_011530362.2
5
1265
XM_011532062.2
XP_011530364.2
5
1265
XM_011532063.2
XP_011530365.2
5
1265
XM_011532064.2
XP_011530366.2
5
1265
XM_011532067.1
XP_011530369.1
5
464
XM_011532070.2
XP_011530372.2
5
1261
XM_011532072.2
XP_011530374.2
5
1261
XM_011532073.2
XP_011530375.2
5
1261
XM_011532075.1
XP_011530377.1
5
444
XM_011532076.2
XP_011530378.1
2
209
XM_017008338.1
XP_016863827.1
5
1266
XM_017008339.1
XP_016863828.1
5
1265
XM_017008340.1
XP_016863829.1
5
899
XM_017008341.1
XP_016863830.1
5
412
XM_017008342.1
XP_016863831.1
3
270
XM_017008343.1
XP_016863832.1
5
464
XM_017008344.1
XP_016863833.1
5
899
XM_017008345.1
XP_016863834.1
5
1261
XM_017008346.1
XP_016863835.1
3
225
XM_017008347.1
XP_016863836.1
5
1261
XM_017008348.1
XP_016863837.1
5
1260
XM_017008350.1
XP_016863839.1
1
49
XM_017008352.1
XP_016863841.1
5
1256
XM_017008353.1
XP_016863842.1
5
899
GenBank mRNA
AK001240.1
-
2
187
AK310421.1
-
5
444
AL137759.1
-
2
197
AL832170.1
-
5
383
BC002840.1
-
5
400
BC018794.1
-
5
399
BC110300.1
-
5
395
DA181086.1
-
5
464
DB230609.1
-
5
421
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
mitophagy in response to mitochondrial depolarization
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0