Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LARP1B SIRNA Assays ›
          Gene Symbol:
          LARP1B
          Gene Name:
          La ribonucleoprotein domain family member 1B
          Gene Aliases:
          LARP2
          Chromosome Location:
          Chr.4: 128060655 - 128222931 on Build GRCh38
          UniGene ID:
          Hs.657067
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s30247

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LARP1B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001278604.1 NP_001265533.1 4, 5 426
          NM_018078.3 NP_060548.2 4, 5 426
          NM_032239.3 NP_115615.2 4, 5 426
          NM_178043.2 NP_835144.1 4, 5 426
          XM_011532057.2 XP_011530359.2 4, 5 1228
          XM_011532058.2 XP_011530360.2 4, 5 1227
          XM_011532059.2 XP_011530361.2 4, 5 1228
          XM_011532060.2 XP_011530362.2 4, 5 1227
          XM_011532062.2 XP_011530364.2 4, 5 1227
          XM_011532063.2 XP_011530365.2 4, 5 1227
          XM_011532064.2 XP_011530366.2 4, 5 1227
          XM_011532067.1 XP_011530369.1 4, 5 426
          XM_011532070.2 XP_011530372.2 4, 5 1223
          XM_011532072.2 XP_011530374.2 4, 5 1223
          XM_011532073.2 XP_011530375.2 4, 5 1223
          XM_011532075.1 XP_011530377.1 4, 5 406
          XM_011532076.2 XP_011530378.1 1, 2 171
          XM_017008338.1 XP_016863827.1 4, 5 1228
          XM_017008339.1 XP_016863828.1 4, 5 1227
          XM_017008340.1 XP_016863829.1 4, 5 861
          XM_017008341.1 XP_016863830.1 4, 5 374
          XM_017008342.1 XP_016863831.1 2, 3 232
          XM_017008343.1 XP_016863832.1 4, 5 426
          XM_017008344.1 XP_016863833.1 4, 5 861
          XM_017008345.1 XP_016863834.1 4, 5 1223
          XM_017008346.1 XP_016863835.1 2, 3 187
          XM_017008347.1 XP_016863836.1 4, 5 1223
          XM_017008348.1 XP_016863837.1 4, 5 1222
          XM_017008350.1 XP_016863839.1 11
          XM_017008352.1 XP_016863841.1 4, 5 1218
          XM_017008353.1 XP_016863842.1 4, 5 861
          GenBank mRNA AK001240.1 - 1, 2 149
          AK310421.1 - 4, 5 406
          AL137759.1 - 1, 2 159
          AL832170.1 - 4, 5 345
          BC002840.1 - 4, 5 362
          BC018794.1 - 4, 5 361
          BC110300.1 - 4, 5 357
          DA181086.1 - 4, 5 426
          DB230609.1 - 4, 5 383

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mitophagy in response to mitochondrial depolarization
          protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0