Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018122.4 | NP_060592.2 | 13, 14 | 1921 | ||
GenBank mRNA | AK001376.1 | - | 9, 10 | 925 | ||
AK022754.1 | - | 9, 10 | 926 | |||
AK290873.1 | - | 13, 14 | 1603 | |||
AL833633.1 | - | 13, 14 | 1606 | |||
BC045173.2 | - | 13, 14 | 1900 | 5273243 |