Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHQ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018130.2 | NP_060600.2 | 2 | 303 | ||
XM_011533895.2 | XP_011532197.1 | 2 | 331 | |||
XM_011533896.2 | XP_011532198.1 | 2 | 331 | |||
XM_011533897.2 | XP_011532199.1 | 2 | 331 | |||
XM_011533898.2 | XP_011532200.1 | 2 | 331 | |||
XM_011533899.1 | XP_011532201.1 | 2 | 331 | |||
XM_017006724.1 | XP_016862213.1 | 2 | 331 | |||
GenBank mRNA | AK001401.1 | - | 2 | 303 | ||
AK092962.1 | - | 2 | 301 | |||
AK296789.1 | - | 2 | 202 | |||
BC017204.2 | - | 2 | 293 | 3617948 | ||
BC017274.2 | - | 2 | 281 | 4840343 | ||
BC025270.1 | - | 2 | 293 | 4622871 | ||
BC032671.1 | - | 2 | 307 | 5538687 | ||
BC039830.1 | - | 2 | 236 | 6064610 | ||
BC047879.2 | - | 2 | 254 | 5541019 |