Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PBRM1 SIRNA Assays ›
Gene Symbol:
PBRM1
Gene Name:
polybromo 1
Gene Aliases:
BAF180, PB1
Chromosome Location:
Chr.3: 52545352 - 52685933 on Build GRCh38
UniGene ID:
Hs.189920
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s30400
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PBRM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_018313.4
NP_060783.3
26
4088
XM_005265279.4
XP_005265336.1
28
4517
XM_005265280.3
XP_005265337.1
27
4334
XM_005265282.3
XP_005265339.1
28
4476
XM_005265283.3
XP_005265340.1
27
4331
XM_011533900.2
XP_011532202.1
30
4683
XM_011533902.2
XP_011532204.1
29
4615
XM_011533903.2
XP_011532205.1
29
4620
XM_017006725.1
XP_016862214.1
29
4590
XM_017006726.1
XP_016862215.1
30
4700
XM_017006727.1
XP_016862216.1
30
4645
XM_017006728.1
XP_016862217.1
29
4641
XM_017006729.1
XP_016862218.1
29
5046
XM_017006730.1
XP_016862219.1
30
4636
XM_017006731.1
XP_016862220.1
29
4619
XM_017006732.1
XP_016862221.1
29
4612
XM_017006733.1
XP_016862222.1
29
5009
XM_017006734.1
XP_016862223.1
29
4582
XM_017006735.1
XP_016862224.1
28
5003
XM_017006736.1
XP_016862225.1
28
5001
XM_017006737.1
XP_016862226.1
28
4923
XM_017006738.1
XP_016862227.1
28
4544
XM_017006739.1
XP_016862228.1
28
4920
XM_017006740.1
XP_016862229.1
28
4461
XM_017006741.1
XP_016862230.1
28
4552
XM_017006742.1
XP_016862231.1
28
4575
XM_017006743.1
XP_016862232.1
29
4974
XM_017006744.1
XP_016862233.1
28
4541
XM_017006745.1
XP_016862234.1
28
4895
XM_017006746.1
XP_016862235.1
28
4892
XM_017006747.1
XP_016862236.1
28
4954
XM_017006748.1
XP_016862237.1
27
4850
XM_017006749.1
XP_016862238.1
27
4416
XM_017006750.1
XP_016862239.1
26
4304
XM_017006751.1
XP_016862240.1
28
4477
XM_017006752.1
XP_016862241.1
29
5049
XM_017006753.1
XP_016862242.1
29
5048
XM_017006754.1
XP_016862243.1
29
4615
XM_017006755.1
XP_016862244.1
28
4552
XM_017006756.1
XP_016862245.1
28
4549
XM_017006757.1
XP_016862246.1
26
4275
XM_017006758.1
XP_016862247.1
26
4406
XM_017006759.1
XP_016862248.1
28
4477
XM_017006760.1
XP_016862249.1
29
4615
XM_017006761.1
XP_016862250.1
29
4624
XM_017006762.1
XP_016862251.1
29
4612
XM_017006763.1
XP_016862252.1
30
4674
XM_017006764.1
XP_016862253.1
28
4552
XM_017006765.1
XP_016862254.1
27
4431
XM_017006766.1
XP_016862255.1
28
4484
GenBank mRNA
AF177387.1
-
25
4133
AF197569.1
-
26
4070
AF225870.1
-
24
4037
AF225871.1
-
24
3918
AF225872.1
-
25
4133
AK001507.1
-
2
160
AY281068.1
-
25
4132
BC015323.1
-
3
424
BC089409.1
-
4
505
BC115009.1
-
25
4145
7962003
BC115010.1
-
25
4074
BC115011.1
-
25
4149
7962010
BC129934.1
-
26
4178
40082629
BC129935.1
-
26
4178
40082630
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
mitotic nuclear division
negative regulation of cell proliferation
covalent chromatin modification
DNA binding
chromatin binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65b4bdbffd-bsnw9:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0