Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SETD5 SIRNA Assays ›
Gene Symbol:
SETD5
Gene Name:
SET domain containing 5
Gene Aliases:
-
Chromosome Location:
Chr.3: 9397700 - 9478154 on Build GRCh38
UniGene ID:
Hs.288164
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s30444
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SETD5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080517.2
NP_001073986.1
20
3669
NM_001292043.1
NP_001278972.1
22
3933
XM_005265301.1
XP_005265358.1
20
3707
XM_005265303.1
XP_005265360.1
23
4145
XM_011533920.1
XP_011532222.1
24
4240
XM_011533921.1
XP_011532223.1
23
3842
XM_011533922.1
XP_011532224.1
22
3803
XM_011533927.1
XP_011532229.1
24
4183
XM_011533928.1
XP_011532230.1
22
3746
XM_011533930.1
XP_011532232.1
24
4184
XM_011533932.1
XP_011532234.1
22
3951
XM_011533933.1
XP_011532235.1
20
3320
XM_017006767.1
XP_016862256.1
24
5902
XM_017006768.1
XP_016862257.1
22
5416
XM_017006769.1
XP_016862258.1
22
7519
XM_017006770.1
XP_016862259.1
22
7501
XM_017006771.1
XP_016862260.1
21
3764
XM_017006772.1
XP_016862261.1
21
7462
XM_017006773.1
XP_016862262.1
22
7462
XM_017006774.1
XP_016862263.1
21
3707
XM_017006775.1
XP_016862264.1
21
7405
XM_017006776.1
XP_016862265.1
23
4047
XM_017006777.1
XP_016862266.1
23
3990
XM_017006778.1
XP_016862267.1
24
4031
XM_017006779.1
XP_016862268.1
22
4008
XM_017006780.1
XP_016862269.1
23
3992
XM_017006781.1
XP_016862270.1
23
3974
XM_017006782.1
XP_016862271.1
23
7558
XM_017006783.1
XP_016862272.1
25
4256
GenBank mRNA
AB051544.1
-
16
3463
AK001569.1
-
9
1852
AK024384.1
-
8
1763
AK025478.1
-
4
777
AK091393.1
-
17
2972
AK127298.1
-
10
2556
AK289529.1
-
11
2251
AL442073.1
-
7
1649
BC020956.1
-
9
1883
3851178
BX648380.1
-
12
2271
CR627408.1
-
19
3285
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-tvqvm:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0