Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPPA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018189.3 | NP_060659.3 | 2 | 153 | ||
XM_011512954.2 | XP_011511256.1 | 2 | 432 | |||
GenBank mRNA | AK001575.1 | - | 2 | 95 | ||
AK022698.1 | - | 2 | 110 | |||
AK290992.1 | - | 2 | 146 | |||
AK298623.1 | - | 2 | 103 | |||
AK299290.1 | - | 2 | 96 | |||
BC032846.2 | - | 2 | 127 | 5267235 |