Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P3H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134418.1 | NP_001127890.1 | 12 | 1497 | ||
NM_018192.3 | NP_060662.2 | 12 | 2130 | |||
XM_011512955.1 | XP_011511257.1 | 12 | 1404 | |||
GenBank mRNA | AJ430351.2 | - | 12 | 1940 | ||
AK001580.1 | - | 12 | 1331 | |||
AK056447.1 | - | 12 | 1940 | |||
AK125134.1 | - | 12 | 1935 | |||
BC005029.1 | - | 12 | 1325 | 4040181 |