Search
Search

RAVER2| Gene Symbol: RAVER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018211.3 | NP_060681.2 | 4 | 881 | ||
| XM_006710738.3 | XP_006710801.2 | 4 | 988 | |||
| XM_011541706.2 | XP_011540008.1 | 4 | 997 | |||
| GenBank mRNA | AB046799.1 | - | 4 | 881 | ||
| AK001632.1 | - | 2 | 44 | |||
| AK299228.1 | - | 2 | 52 | |||
| BC065303.1 | - | 2 | 272 | |||