Search
Search

SLC35C1
LOC105747689| Gene Symbol: SLC35C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145265.1 | NP_001138737.1 | 2 | 817 | ||
| NM_001145266.1 | NP_001138738.1 | 2 | 720 | |||
| NM_018389.4 | NP_060859.4 | 1 | 1091 | |||
| XM_011520203.2 | XP_011518505.1 | 1 | 1091 | |||
| GenBank mRNA | AF323970.1 | - | 1 | 379 | ||
| AF326199.1 | - | 1 | 988 | |||
| AK002182.1 | - | 2 | 493 | |||
| AK027394.1 | - | 1 | 1068 | |||
| AK027747.1 | - | 1 | 1032 | |||
| AK074929.1 | - | 1 | 1092 | |||
| AK091245.1 | - | 1 | 1775 | |||
| AK225428.1 | - | 2 | 515 | |||
| AK315473.1 | - | 1 | 793 | |||
| BC001427.2 | - | 1 | 396 | 3139599 | ||
| Gene Symbol: LOC105747689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK130181.1 | - | 539 | |||