Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF331 SIRNA Assays ›
Gene Symbol:
ZNF331
Gene Name:
zinc finger protein 331
Gene Aliases:
RITA, ZNF361, ZNF463
Chromosome Location:
Chr.19: 53503819 - 53580269 on Build GRCh38
UniGene ID:
Hs.185674
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s30859
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF331
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001079906.1
NP_001073375.1
6
1036
NM_001079907.1
NP_001073376.1
6
1056
NM_001253798.1
NP_001240727.1
7
1054
NM_001253799.1
NP_001240728.1
6
1024
NM_001253800.1
NP_001240729.1
6
983
NM_001253801.2
NP_001240730.1
5
946
NM_001317113.1
NP_001304042.1
5
972
NM_001317114.1
NP_001304043.1
6
889
NM_001317115.1
NP_001304044.1
7
1066
NM_001317116.1
NP_001304045.1
5
1027
NM_001317117.1
NP_001304046.1
6
976
NM_001317118.1
NP_001304047.1
6
1057
NM_001317119.1
NP_001304048.1
5
1026
NM_001317120.1
NP_001304049.1
7
2010
NM_001317121.1
NP_001304050.1
5
930
NM_018555.5
NP_061025.5
7
2048
XM_011527076.2
XP_011525378.1
8
2567
XM_011527078.2
XP_011525380.1
8
2166
XM_017026936.1
XP_016882425.1
12
2760
XM_017026937.1
XP_016882426.1
12
2285
XM_017026938.1
XP_016882427.1
7
1279
XM_017026939.1
XP_016882428.1
6
1051
XM_017026940.1
XP_016882429.1
5
931
GenBank mRNA
AF251515.2
-
6
990
AF272148.1
-
5
800
AK075226.1
-
6
957
AK128736.1
-
7
1934
AK312889.1
-
5
909
BC009365.2
-
5
773
BC009433.1
-
8
1273
3355780
CR749560.1
-
7
1958
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
RNA polymerase II core promoter proximal region sequence-specific DNA binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0