Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC42BPG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017525.2 | NP_059995.2 | 21 | 2434 | ||
XM_011545155.2 | XP_011543457.1 | 21 | 2549 | |||
XM_011545156.2 | XP_011543458.1 | 21 | 2529 | |||
XM_011545157.2 | XP_011543459.1 | 21 | 2550 | |||
XM_011545158.2 | XP_011543460.1 | 21 | 2550 | |||
XM_011545159.2 | XP_011543461.1 | 21 | 2550 | |||
XM_011545160.2 | XP_011543462.1 | 22 | 2501 | |||
XM_011545161.2 | XP_011543463.1 | 21 | 2550 | |||
XM_017017996.1 | XP_016873485.1 | 21 | 2549 | |||
XM_017017997.1 | XP_016873486.1 | 21 | 2550 | |||
XM_017017998.1 | XP_016873487.1 | 21 | 2550 | |||
XM_017017999.1 | XP_016873488.1 | 11 | 1129 | |||
GenBank mRNA | AY648038.1 | - | 21 | 2434 |