Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KIF27 SIRNA Assays ›
Gene Symbol:
KIF27
Gene Name:
kinesin family member 27
Gene Aliases:
-
Chromosome Location:
Chr.9: 83832795 - 83921539 on Build GRCh38
UniGene ID:
Hs.697514
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s31013
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KIF27
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001271927.1
NP_001258856.1
3
652
NM_001271928.1
NP_001258857.1
3
652
NM_017576.2
NP_060046.1
3
652
XM_011518848.2
XP_011517150.1
3
653
XM_011518849.2
XP_011517151.1
4
769
XM_011518850.2
XP_011517152.1
3
742
XM_011518854.2
XP_011517156.1
4
872
XM_011518856.2
XP_011517158.1
4
872
XM_011518857.2
XP_011517159.1
4
849
XM_011518858.2
XP_011517160.1
1
85
XM_017014900.1
XP_016870389.1
5
921
XM_017014901.1
XP_016870390.1
5
803
XM_017014902.1
XP_016870391.1
4
849
XM_017014903.1
XP_016870392.1
4
872
XM_017014904.1
XP_016870393.1
4
769
XM_017014905.1
XP_016870394.1
4
872
XM_017014906.1
XP_016870395.1
4
872
XM_017014907.1
XP_016870396.1
4
872
XM_017014908.1
XP_016870397.1
4
872
XM_017014909.1
XP_016870398.1
3
474
XM_017014910.1
XP_016870399.1
4
769
XM_017014911.1
XP_016870400.1
2
557
XM_017014912.1
XP_016870401.1
3
650
XM_017014913.1
XP_016870402.1
4
842
XM_017014914.1
XP_016870403.1
3
726
XM_017014915.1
XP_016870404.1
4
812
XM_017014916.1
XP_016870405.1
3
712
GenBank mRNA
AK297689.1
-
3
609
AY237536.1
-
3
614
AY237537.1
-
3
614
AY237538.1
-
3
614
BC140788.1
-
3
613
9021656
Back To Top
More Information
Panther Classification:
Molecular Function -
microtubule binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
epithelial cilium movement
microtubule-based movement
ventricular system development
cilium assembly
microtubule motor activity
ATP binding
microtubule binding
ATPase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-kz7s5:80/10.239.165.170:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0