Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SYBU SIRNA Assays ›
          Gene Symbol:
          SYBU
          Gene Name:
          syntabulin
          Gene Aliases:
          GOLSYN, OCSYN, SNPHL
          Chromosome Location:
          Chr.8: 109574176 - 109691791 on Build GRCh38
          UniGene ID:
          Hs.390738
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s31144

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SYBU
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001099743.1 NP_001093213.1 8 2131
          NM_001099744.1 NP_001093214.1 8 2134
          NM_001099745.1 NP_001093215.1 8 1970
          NM_001099746.1 NP_001093216.1 6 1684
          NM_001099747.1 NP_001093217.1 8 1888
          NM_001099748.1 NP_001093218.1 8 1891
          NM_001099749.1 NP_001093219.1 6 1605
          NM_001099750.1 NP_001093220.1 9 2012
          NM_001099751.1 NP_001093221.1 8 1977
          NM_001099752.1 NP_001093222.1 8 1980
          NM_001099753.1 NP_001093223.1 7 1778
          NM_001099754.1 NP_001093224.1 7 1781
          NM_001099755.1 NP_001093225.1 6 1576
          NM_001099756.1 NP_001093226.1 6 1852
          NM_017786.5 NP_060256.3 9 2009
          XM_005250999.1 XP_005251056.1 5 1280
          XM_005251000.1 XP_005251057.1 5 1289
          XM_011517154.2 XP_011515456.1 4 4039
          XM_017013614.1 XP_016869103.1 7 1700
          XM_017013615.1 XP_016869104.1 7 1674
          GenBank mRNA AB040905.1 - 6 1434
          AB195676.1 - 9 1942
          AB195677.1 - 8 1821
          AB195678.1 - 6 1538
          AB195679.1 - 6 2337
          AK000373.1 - 6 1663
          AK055246.1 - 8 1901
          AK090559.1 - 6 1571
          AK091374.1 - 3 1624
          AK096558.1 - 7 1776
          AK290469.1 - 8 1970
          AK297182.1 - 5 1274
          AL713645.1 - 9 1903
          AL834406.1 - 3 1049
          AY705385.1 - 7 1618
          BC012082.1 - 6 1551 4661631

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          SNARE protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of insulin secretion involved in cellular response to glucose stimulus
          protein binding
          microtubule binding
          syntaxin-1 binding
          kinesin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0