Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018060.3 | NP_060530.3 | 9 | 1271 | ||
GenBank mRNA | AK001188.1 | - | 7 | 746 | ||
AK001726.1 | - | 3 | 260 | |||
AK291432.1 | - | 9 | 1080 | |||
AY267462.1 | - | 9 | 1282 | |||
BC010218.1 | - | 5 | 451 | |||
BC040376.1 | - | 9 | 1004 | |||
BC047880.1 | - | 8 | 798 | |||
BC137438.1 | - | 9 | 1241 | 9021436 | ||
D28500.1 | - | 9 | 1112 |