Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145159.2 | NP_001138631.1 | 2 | 345 | ||
NM_001172562.1 | NP_001166033.1 | 2 | 111 | |||
NM_018250.3 | NP_060720.2 | 2 | 345 | |||
XM_006716356.2 | XP_006716419.1 | 2 | 142 | |||
XM_006716357.3 | XP_006716420.1 | 2 | 142 | |||
XM_011544573.1 | XP_011542875.1 | 2 | 142 | |||
XM_011544574.1 | XP_011542876.1 | 1 | 43 | |||
XM_011544575.2 | XP_011542877.1 | 3 | 563 | |||
XM_011544576.1 | XP_011542878.1 | 2 | 142 | |||
GenBank mRNA | AK001733.1 | - | 2 | 126 | ||
AK225072.1 | - | 2 | 78 | |||
AK225530.1 | - | 2 | 142 | |||
AK295977.1 | - | 2 | 107 | |||
AK298468.1 | - | 2 | 111 | |||
AK300593.1 | - | 2 | 105 | |||
BC016687.1 | - | 2 | 77 | |||
BC025267.1 | - | 2 | 95 | 5426533 | ||
DC339488.1 | - | 2 | 346 |