Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MBNL3 SIRNA Assays ›
Gene Symbol:
MBNL3
Gene Name:
muscleblind like splicing regulator 3
Gene Aliases:
CHCR, MBLX, MBLX39, MBXL
Chromosome Location:
Chr.X: 132369314 - 132490030 on Build GRCh38
UniGene ID:
Hs.105134
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s31535
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MBNL3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170701.1
NP_001164172.1
3
432
NM_001170702.1
NP_001164173.1
3
432
NM_001170703.1
NP_001164174.1
3
365
NM_001170704.1
NP_001164175.1
4
632
NM_018388.3
NP_060858.2
3
595
NM_133486.2
NP_597846.1
3
595
XM_005262431.3
XP_005262488.1
4
1407
XM_005262434.3
XP_005262491.1
4
1402
XM_005262436.3
XP_005262493.1
3
1295
XM_005262437.3
XP_005262494.1
3
433
XM_005262438.3
XP_005262495.1
3
433
XM_005262439.3
XP_005262496.1
3
365
XM_005262440.3
XP_005262497.1
5
1452
XM_005262441.3
XP_005262498.1
3
556
XM_005262442.3
XP_005262499.1
3
527
XM_011531364.2
XP_011529666.1
2
1443
XM_017029632.1
XP_016885121.1
4
8837
XM_017029633.1
XP_016885122.1
5
1572
XM_017029634.1
XP_016885123.1
4
8837
XM_017029635.1
XP_016885124.1
4
8837
XM_017029636.1
XP_016885125.1
4
1336
XM_017029637.1
XP_016885126.1
4
8837
XM_017029638.1
XP_016885127.1
5
1470
XM_017029639.1
XP_016885128.1
3
545
GenBank mRNA
AB077698.1
-
3
595
AB077699.1
-
3
595
AF467070.1
-
3
595
AF491305.1
-
3
516
AJ427918.1
-
3
595
AJ427919.1
-
3
595
AK002178.1
-
3
401
AK300248.1
-
3
364
AL133625.1
-
2
340
AY072692.1
-
3
595
AY372211.1
-
3
366
BC042090.1
-
4
631
5761647
BC074775.2
-
3
558
30915333
BC074776.2
-
3
558
30915555
BU569439.1
-
3
435
DB023330.1
-
3
365
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
mRNA processing
multicellular organism development
RNA splicing
regulation of RNA splicing
negative regulation of myoblast differentiation
protein binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-xq6xf:80/100.66.76.183:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0