Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146688.1 | NP_001140160.1 | 5 | 535 | ||
NM_018433.5 | NP_060903.2 | 5 | 804 | |||
XM_006712051.3 | XP_006712114.1 | 4 | 507 | |||
XM_017004493.1 | XP_016859982.1 | 5 | 823 | |||
XM_017004494.1 | XP_016859983.1 | 5 | 593 | |||
GenBank mRNA | AB018285.2 | - | 5 | 567 | ||
AK304276.1 | - | 3 | 321 | |||
AL832150.1 | - | 5 | 570 | |||
BC038297.2 | - | 5 | 581 | |||
BG723267.1 | - | 5 | 587 | |||
BX640698.1 | - | 5 | 605 | |||
BX647271.1 | - | 5 | 573 | |||
BX647752.1 | - | 5 | 633 | |||
BX648841.1 | - | 5 | 536 | |||
BX649109.1 | - | 5 | 574 | |||
CR749581.1 | - | 5 | 572 |