Search
Search

GLT8D1| Gene Symbol: GLT8D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010983.2 | NP_001010983.1 | 11 | 1923 | ||
| NM_001278280.1 | NP_001265209.1 | 11 | 1919 | |||
| NM_001278281.1 | NP_001265210.1 | 11 | 1667 | |||
| NM_018446.3 | NP_060916.1 | 10 | 1694 | |||
| NM_152932.2 | NP_690909.1 | 10 | 1252 | |||
| XM_017006857.1 | XP_016862346.1 | 12 | 2082 | |||
| XM_017006858.1 | XP_016862347.1 | 11 | 1853 | |||
| XM_017006859.1 | XP_016862348.1 | 10 | 1402 | |||
| GenBank mRNA | AF157318.1 | - | 10 | 1139 | ||
| AF175227.1 | - | 11 | 1410 | |||
| AF176777.1 | - | 11 | 1415 | |||
| AJ271710.1 | - | 9 | 1021 | |||
| AK027517.1 | - | 10 | 1160 | |||
| AK075031.1 | - | 7 | 1674 | |||
| AY358579.1 | - | 10 | 1173 | |||
| BC119670.1 | - | 10 | 1138 | 40116196 | ||
| BC119671.1 | - | 10 | 1138 | 40116197 | ||
| CR749821.1 | - | 11 | 1376 | |||