Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018728.3 | NP_061198.2 | 17 | 2203 | ||
XM_011521781.2 | XP_011520083.1 | 17 | 2203 | |||
XM_017022408.1 | XP_016877897.1 | 16 | 2068 | |||
XM_017022409.1 | XP_016877898.1 | 12 | 1510 | |||
XM_017022410.1 | XP_016877899.1 | 16 | 2053 | |||
GenBank mRNA | AF272390.1 | - | 17 | 2060 | ||
AK304136.1 | - | 18 | 2318 | |||
BC144611.1 | - | 17 | 1988 |