Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022052.1 | NP_071335.1 | 3 | 307 | ||
GenBank mRNA | AF346619.1 | - | 3 | 307 | ||
AJ277527.1 | - | 3 | 307 | |||
AK302447.1 | - | 3 | 308 | |||
AK302586.1 | - | 2 | 273 | |||
BC031616.1 | - | 3 | 300 | 5170537 |