Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMEM234 SIRNA Assays ›
Gene Symbol:
TMEM234
Gene Name:
transmembrane protein 234
Gene Aliases:
AASL548, C1orf91, PRO1105, RP4-622L5, dJ622L5.7
Chromosome Location:
Chr.1: 32204807 - 32222371 on Build GRCh38
UniGene ID:
Hs.272299
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s31874
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMEM234
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_019118.4
NP_061991.3
3
259
NR_133634.1
3
259
NR_133635.1
3
259
NR_133636.1
3
259
XM_011541795.2
XP_011540097.1
3
241
XM_017001812.1
XP_016857301.1
3
241
XM_017001813.1
XP_016857302.1
3
241
XM_017001814.1
XP_016857303.1
3
241
XM_017001815.1
XP_016857304.1
3
241
XM_017001816.1
XP_016857305.1
3
241
XM_017001817.1
XP_016857306.1
3
241
XM_017001818.1
XP_016857307.1
3
241
XM_017001819.1
XP_016857308.1
3
241
XM_017001820.1
XP_016857309.1
3
241
XM_017001821.1
XP_016857310.1
3
241
XM_017001822.1
XP_016857311.1
3
241
XM_017001823.1
XP_016857312.1
3
241
XM_017001824.1
XP_016857313.1
3
241
XM_017001825.1
XP_016857314.1
3
241
XM_017001826.1
XP_016857315.1
3
241
XM_017001827.1
XP_016857316.1
3
241
XM_017001828.1
XP_016857317.1
3
241
XM_017001829.1
XP_016857318.1
3
241
XM_017001830.1
XP_016857319.1
3
241
XM_017001831.1
XP_016857320.1
2
101
GenBank mRNA
AF258548.1
-
3
236
AK295232.1
-
3
259
AK312046.1
-
3
234
AY358586.1
-
3
237
BC001256.1
-
3
221
BC004885.2
-
3
213
3837394
BC038842.1
-
3
236
5742036
BP307734.1
-
3
229
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0