Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YLPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019589.2 | NP_062535.2 | 7, 8 | 5065 | ||
XM_005267860.3 | XP_005267917.1 | 7, 8 | 5109 | |||
XM_011536966.2 | XP_011535268.1 | 8, 9 | 4224 | |||
XM_011536967.1 | XP_011535269.1 | 6, 7 | 2991 | |||
GenBank mRNA | AK090435.1 | - | 5, 6 | 3821 | ||
AK297582.1 | - | 3, 4 | 636 | |||
AL832365.1 | - | 4, 5 | 3600 | |||
L40403.1 | - | 1, 2 | 168 |