Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KYAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008661.2 | NP_001008661.1 | 11 | 1434 | ||
NM_001008662.2 | NP_001008662.1 | 10 | 1334 | |||
GenBank mRNA | AF091090.1 | - | 7 | 637 | ||
AK057176.1 | - | 10 | 1150 | |||
AK301326.1 | - | 11 | 1121 | |||
AK315917.1 | - | 11 | 1131 | |||
AL833593.1 | - | 12 | 1236 | |||
AY028624.1 | - | 11 | 1254 | |||
BC000819.1 | - | 8 | 807 | 3453829 |