Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CELF4 SIRNA Assays ›
Gene Symbol:
CELF4
Gene Name:
CUGBP, Elav-like family member 4
Gene Aliases:
BRUNOL4, CELF-4
Chromosome Location:
Chr.18: 37243040 - 37566052 on Build GRCh38
UniGene ID:
Hs.435976
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s32298
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CELF4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025087.1
NP_001020258.1
1
532
NM_001025088.1
NP_001020259.1
1
532
NM_001025089.1
NP_001020260.1
1
532
NM_020180.3
NP_064565.1
1
532
XM_005258308.3
XP_005258365.1
1
544
XM_011526079.2
XP_011524381.1
1
546
XM_011526081.2
XP_011524383.1
1
383
XM_011526083.2
XP_011524385.1
1
546
XM_011526084.2
XP_011524386.1
1
546
XM_011526085.2
XP_011524387.1
1
547
XM_011526086.2
XP_011524388.1
1
545
XM_011526087.2
XP_011524389.1
1
547
XM_011526090.2
XP_011524392.1
1
544
XM_011526091.2
XP_011524393.1
1
544
XM_011526092.2
XP_011524394.1
1
544
XM_017025840.1
XP_016881329.1
1
409
XM_017025841.1
XP_016881330.1
1
543
XM_017025842.1
XP_016881331.1
1
545
XM_017025843.1
XP_016881332.1
1
545
XM_017025844.1
XP_016881333.1
1
545
XM_017025845.1
XP_016881334.1
1
545
XM_017025846.1
XP_016881335.1
1
543
XM_017025847.1
XP_016881336.1
1
543
XM_017025848.1
XP_016881337.1
1
544
XM_017025849.1
XP_016881338.1
1
544
XM_017025850.1
XP_016881339.1
1
409
XM_017025851.1
XP_016881340.1
1
409
XM_017025852.1
XP_016881341.1
1
409
XM_017025853.1
XP_016881342.1
1
545
XM_017025854.1
XP_016881343.1
1
545
XM_017025855.1
XP_016881344.1
1
545
XM_017025856.1
XP_016881345.1
1
545
XM_017025857.1
XP_016881346.1
1
546
XM_017025858.1
XP_016881347.1
1
383
XM_017025859.1
XP_016881348.1
1
383
XM_017025860.1
XP_016881349.1
1
544
XM_017025861.1
XP_016881350.1
1
409
XM_017025862.1
XP_016881351.1
1
409
XM_017025863.1
XP_016881352.1
1
391
XM_017025864.1
XP_016881353.1
1
383
XM_017025865.1
XP_016881354.1
1
409
GenBank mRNA
AB209774.1
-
1
326
AF248650.1
-
1
271
AF329265.1
-
1
136
AK074596.1
-
1
264
AK295009.1
-
1
146
BC001946.1
-
1
293
2820541
BC004167.2
-
1
293
2820541
BC045711.1
-
1
219
5261155
BQ637723.1
-
1
539
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001174074, NM_133195, NM_001146294, NM_001146292, NM_001146295, and others.
Gene Ontology Categories:
Function(s)
Process(es)
alternative mRNA splicing, via spliceosome
regulation of alternative mRNA splicing, via spliceosome
mRNA splice site selection
germ cell development
embryo development
negative regulation of translation
negative regulation of mRNA splicing, via spliceosome
positive regulation of mRNA splicing, via spliceosome
negative regulation of excitatory postsynaptic potential
nucleotide binding
translation repressor activity, nucleic acid binding
mRNA binding
BRE binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0