Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CELF4 SIRNA Assays ›
          Gene Symbol:
          CELF4
          Gene Name:
          CUGBP, Elav-like family member 4
          Gene Aliases:
          BRUNOL4, CELF-4
          Chromosome Location:
          Chr.18: 37243040 - 37566052 on Build GRCh38
          UniGene ID:
          Hs.435976
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s32300

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CELF4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001025087.1 NP_001020258.1 1 649
          NM_001025088.1 NP_001020259.1 1 649
          NM_001025089.1 NP_001020260.1 1 649
          NM_020180.3 NP_064565.1 1 649
          XM_005258308.3 XP_005258365.1 1 661
          XM_011526079.2 XP_011524381.1 1 663
          XM_011526081.2 XP_011524383.1 1 500
          XM_011526083.2 XP_011524385.1 1 663
          XM_011526084.2 XP_011524386.1 1 663
          XM_011526085.2 XP_011524387.1 1 664
          XM_011526086.2 XP_011524388.1 1 662
          XM_011526087.2 XP_011524389.1 1 664
          XM_011526090.2 XP_011524392.1 1 661
          XM_011526091.2 XP_011524393.1 1 661
          XM_011526092.2 XP_011524394.1 1 661
          XM_017025840.1 XP_016881329.1 1 526
          XM_017025841.1 XP_016881330.1 1 660
          XM_017025842.1 XP_016881331.1 1 662
          XM_017025843.1 XP_016881332.1 1 662
          XM_017025844.1 XP_016881333.1 1 662
          XM_017025845.1 XP_016881334.1 1 662
          XM_017025846.1 XP_016881335.1 1 660
          XM_017025847.1 XP_016881336.1 1 660
          XM_017025848.1 XP_016881337.1 1 661
          XM_017025849.1 XP_016881338.1 1 661
          XM_017025850.1 XP_016881339.1 1 526
          XM_017025851.1 XP_016881340.1 1 526
          XM_017025852.1 XP_016881341.1 1 526
          XM_017025853.1 XP_016881342.1 1 662
          XM_017025854.1 XP_016881343.1 1 662
          XM_017025855.1 XP_016881344.1 1 662
          XM_017025856.1 XP_016881345.1 1 662
          XM_017025857.1 XP_016881346.1 1 663
          XM_017025858.1 XP_016881347.1 1 500
          XM_017025859.1 XP_016881348.1 1 500
          XM_017025860.1 XP_016881349.1 1 661
          XM_017025861.1 XP_016881350.1 1 526
          XM_017025862.1 XP_016881351.1 1 526
          XM_017025863.1 XP_016881352.1 1 508
          XM_017025864.1 XP_016881353.1 1 500
          XM_017025865.1 XP_016881354.1 1 526
          GenBank mRNA AB209774.1 - 1 443
          AF248650.1 - 1 388
          AF329265.1 - 1 253
          AK074596.1 - 1 381
          AK295009.1 - 1 263
          BC001946.1 - 1 410 2820541
          BC004167.2 - 1 410 2820541
          BC045711.1 - 1 336 5261155

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001107400.
          This siRNA also targets the following transcripts in Mouse: NM_001174074, NM_133195, NM_001146294, NM_001146292, NM_001146295, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          alternative mRNA splicing, via spliceosome
          regulation of alternative mRNA splicing, via spliceosome
          mRNA splice site selection
          germ cell development
          embryo development
          negative regulation of translation
          negative regulation of mRNA splicing, via spliceosome
          positive regulation of mRNA splicing, via spliceosome
          negative regulation of excitatory postsynaptic potential
          nucleotide binding
          translation repressor activity, nucleic acid binding
          mRNA binding
          BRE binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0