Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other AGPAT3 SIRNA Assays ›
Gene Symbol:
AGPAT3
Gene Name:
1-acylglycerol-3-phosphate O-acyltransferase 3
Gene Aliases:
LPAAT-GAMMA1, LPAAT3
Chromosome Location:
Chr.21: 43865160 - 43987594 on Build GRCh38
UniGene ID:
Hs.248785
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s32329
Validated
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
7,898.00
Your Price
Online offer:
7898.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: AGPAT3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001037553.1
NP_001032642.1
5, 6
777
NM_020132.4
NP_064517.1
6, 7
897
XM_005261160.4
XP_005261217.1
6, 7
898
XM_006724029.3
XP_006724092.1
5, 6
826
XM_006724030.3
XP_006724093.1
5, 6
794
XM_006724031.3
XP_006724094.1
5, 6
797
XM_011529660.2
XP_011527962.1
5, 6
636
XM_011529661.2
XP_011527963.1
5, 6
951
XM_011529662.2
XP_011527964.1
7, 8
1601
XM_011529663.2
XP_011527965.1
5, 6
810
XM_011529664.2
XP_011527966.1
5, 6
751
XM_011529665.2
XP_011527967.1
7, 8
1252
XM_017028408.1
XP_016883897.1
5, 6
780
XM_017028409.1
XP_016883898.1
5, 6
858
XM_017028410.1
XP_016883899.1
5, 6
863
XM_017028411.1
XP_016883900.1
5, 6
736
XM_017028412.1
XP_016883901.1
6, 7
1829
XM_017028413.1
XP_016883902.1
6, 7
797
GenBank mRNA
AB040138.1
-
5, 6
852
AF156774.1
-
6, 7
858
AF156775.1
-
5, 6
721
AK125804.1
-
3, 4
3514
AL832919.1
-
3, 4
469
AY358704.1
-
946
BC011971.1
-
6, 7
871
4148915
BC040603.1
-
7, 8
1601
5270905
BC063552.1
-
5, 6
1167
4800514
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00220017_m1
Panther Classification:
Molecular Function -
acyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
phosphatidic acid biosynthetic process
phospholipid biosynthetic process
CDP-diacylglycerol biosynthetic process
triglyceride biosynthetic process
1-acylglycerol-3-phosphate O-acyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0