Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANKS1B SIRNA Assays ›
          Gene Symbol:
          ANKS1B
          Gene Name:
          ankyrin repeat and sterile alpha motif domain containing 1B
          Gene Aliases:
          AIDA, AIDA-1, ANKS2, EB-1, EB1, cajalin-2
          Chromosome Location:
          Chr.12: 98729904 - 99984727 on Build GRCh38
          UniGene ID:
          Hs.506458
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s32344

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ANKS1B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001204065.1 NP_001190994.1 6, 8 836
          NM_001204066.1 NP_001190995.1 7, 9 703
          NM_001204067.1 NP_001190996.1 7, 9 797
          NM_001204068.1 NP_001190997.1 7, 9 1260
          NM_001204069.1 NP_001190998.1 7, 9 1152
          NM_001204070.1 NP_001190999.1 6, 8 1080
          NM_001204079.1 NP_001191008.1 6, 8 839
          NM_001204080.1 NP_001191009.1 5, 7 764
          NM_001204081.1 NP_001191010.1 5, 7 764
          NM_020140.3 NP_064525.1 6, 8 1080
          NM_152788.4 NP_690001.3 21, 23 3692
          NM_181670.3 NP_858056.2 8, 10 1332
          XM_005269028.4 XP_005269085.1 21, 23 3303
          XM_005269029.4 XP_005269086.1 21, 23 3263
          XM_005269032.3 XP_005269089.1 5, 7 628
          XM_006719504.3 XP_006719567.1 22, 24 3837
          XM_006719505.3 XP_006719568.1 22, 24 3840
          XM_006719506.3 XP_006719569.1 22, 24 3837
          XM_006719507.3 XP_006719570.1 22, 24 3840
          XM_006719508.3 XP_006719571.1 22, 24 3840
          XM_006719509.3 XP_006719572.1 21, 23 3765
          XM_006719510.3 XP_006719573.1 21, 23 3765
          XM_006719512.3 XP_006719575.1 20, 22 3585
          XM_006719513.3 XP_006719576.1 20, 22 3585
          XM_006719514.3 XP_006719577.1 20, 22 3573
          XM_011538571.2 XP_011536873.1 22, 24 3738
          XM_017019651.1 XP_016875140.1 22, 24 3840
          XM_017019652.1 XP_016875141.1 22, 24 3828
          XM_017019653.1 XP_016875142.1 22, 24 3837
          XM_017019654.1 XP_016875143.1 22, 24 3825
          XM_017019655.1 XP_016875144.1 22, 24 3837
          XM_017019656.1 XP_016875145.1 22, 24 3735
          XM_017019657.1 XP_016875146.1 21, 23 3663
          XM_017019658.1 XP_016875147.1 20, 22 3585
          XM_017019659.1 XP_016875148.1 8, 10 1331
          XM_017019660.1 XP_016875149.1 7, 9 1256
          XM_017019661.1 XP_016875150.1 9, 11 1042
          XM_017019662.1 XP_016875151.1 9, 11 1175
          XM_017019663.1 XP_016875152.1 8, 10 1226
          XM_017019664.1 XP_016875153.1 8, 10 912
          XM_017019665.1 XP_016875154.1 6, 8 1076
          GenBank mRNA AF145204.1 - 15, 17 2396
          AF164792.1 - 6, 8 1013
          AK289768.1 - 8, 10 1297
          AK294191.1 - 7, 9 797
          AK294994.1 - 5, 7 636
          AK295988.1 - 6, 8 809
          AK315954.1 - 6, 8 691
          AM980945.1 - 6, 8 695
          AY281131.1 - 21, 23 3278
          AY281132.1 - 7, 9 953
          AY283057.2 - 13, 15 2129
          AY356353.1 - 5, 7 593
          AY620824.1 - 12, 14 2003
          AY753193.1 - 6, 8 773
          BC026313.1 - 8, 10 1227
          BC068451.1 - 7, 9 703
          BC142669.1 - 6, 8 1027
          BC150204.1 - 7, 9 1099

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001271371, XM_006241248, XM_006241249, XM_008765241, XM_008765243, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001128086, NM_001177397, NM_181398, NM_001177396, NM_001177398, and others.

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of synaptic plasticity by receptor localization to synapse
          ephrin receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0