Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARNTL2 SIRNA Assays ›
Gene Symbol:
ARNTL2
Gene Name:
aryl hydrocarbon receptor nuclear translocator like 2
Gene Aliases:
BMAL2, CLIF, MOP9, PASD9, bHLHe6
Chromosome Location:
Chr.12: 27332854 - 27425813 on Build GRCh38
UniGene ID:
Hs.445447
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s32435
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARNTL2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001248002.1
NP_001234931.1
9, 10
1348
NM_001248003.1
NP_001234932.1
8, 9
1279
NM_001248004.1
NP_001234933.1
8, 9
1246
NM_001248005.1
NP_001234934.1
8, 9
1279
NM_020183.4
NP_064568.3
10, 11
1390
XM_006719112.3
XP_006719175.1
9, 10
1349
XM_006719114.3
XP_006719177.1
9, 10
1080
XM_011520766.2
XP_011519068.1
10, 11
1391
XM_011520768.2
XP_011519070.1
9, 10
1299
XM_011520769.2
XP_011519071.1
9, 10
1289
XM_011520770.2
XP_011519072.1
10, 11
1209
XM_011520771.2
XP_011519073.1
10, 11
1175
XM_017019666.1
XP_016875155.1
7, 8
1120
XM_017019667.1
XP_016875156.1
10, 11
1407
XM_017019668.1
XP_016875157.1
10, 11
1204
XM_017019669.1
XP_016875158.1
9, 10
1051
GenBank mRNA
AB039921.1
-
7, 8
922
AF231338.1
-
8, 9
1075
AF231339.1
-
7, 8
1036
AF246960.1
-
10, 11
1189
AF246961.1
-
9, 10
1147
AF246962.1
-
8, 9
1078
AF246963.1
-
8, 9
1045
AF256215.1
-
1288
AK296706.1
-
8, 9
1213
BC000172.3
-
7, 8
997
BC125061.1
-
8, 9
1040
40117267
BC125062.1
-
8, 9
1078
40117268
Back To Top
More Information
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
circadian rhythm
entrainment of circadian clock
positive regulation of circadian rhythm
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
protein dimerization activity
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0