Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193557.1 | NP_001180486.1 | 2 | 119 | ||
NM_020189.5 | NP_064574.1 | 2 | 191 | |||
NR_036471.1 | 2 | 191 | ||||
NR_036472.1 | 2 | 191 | ||||
GenBank mRNA | AF173296.1 | - | 2 | 261 | ||
AF201940.1 | - | 2 | 206 | |||
AK000487.1 | - | 2 | 95 | |||
AK095651.1 | - | 2 | 190 | |||
AK290752.1 | - | 2 | 135 | |||
AK316560.1 | - | 2 | 186 | |||
BC007870.2 | - | 2 | 107 | 4303290 | ||
CR457183.1 | - | 1 | 45 | |||
DA548665.1 | - | 2 | 65 | |||
DA762166.1 | - | 2 | 173 | |||
DB168034.1 | - | 2 | 191 |