Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DTWD1 SIRNA Assays ›
          Gene Symbol:
          DTWD1 NMNAT3
          Gene Name:
          DTW domain containing 1
          nicotinamide nucleotide adenylyltransferase 3
          Gene Aliases:
          MDS009
          FKSG76, PNAT-3, PNAT3
          Chromosome Location:
          Chr.15: 49620980 - 49654448 on Build GRCh38
          Chromosome Location:
          Chr.3: 139560180 - 139678050 on Build GRCh38
          UniGene ID:
          Hs.127432
          Hs.208673
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s32537

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DTWD1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001144955.1 NP_001138427.1 4 584
          NM_020234.5 NP_064619.2 5 712
          XM_011521815.1 XP_011520117.1 4 633
          XM_017022419.1 XP_016877908.1 4 631
          XM_017022420.1 XP_016877909.1 5 1493
          XM_017022421.1 XP_016877910.1 5 724
          XM_017022422.1 XP_016877911.1 4 1365
          XM_017022423.1 XP_016877912.1 4 633
          XM_017022424.1 XP_016877913.1 5 704
          XM_017022425.1 XP_016877914.1 5 704
          XM_017022426.1 XP_016877915.1 4 1365
          XM_017022427.1 XP_016877916.1 5 649
          XM_017022428.1 XP_016877917.1 5 654
          XM_017022429.1 XP_016877918.1 5 1600
          XM_017022430.1 XP_016877919.1 5 803
          XM_017022431.1 XP_016877920.1 4 245
          GenBank mRNA AF168717.1 - 4 562
          AK290184.1 - 4 553
          BC032535.2 - 5 652 5527713
          BC093073.1 - 5 642 30326731
          Gene Symbol: NMNAT3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001320510.1 NP_001307439.1 4 645
          XM_017006328.1 XP_016861817.1 5 662
          XM_017006329.1 XP_016861818.1 5 659
          XM_017006330.1 XP_016861819.1 4 622
          XM_017006331.1 XP_016861820.1 5 735
          XM_017006332.1 XP_016861821.1 5 4676
          XM_017006333.1 XP_016861822.1 3 525
          XM_017006334.1 XP_016861823.1 3 522
          XM_017006339.1 XP_016861828.1 4 560
          XM_017006340.1 XP_016861829.1 3 529

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001013921, XM_008762154, XM_006234910, XM_006234914, XM_006234915, and others.

          Panther Classification:

          Molecular Function -

          nucleotidyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          NAD biosynthetic process
          response to wounding
          NAD metabolic process
          'de novo' NAD biosynthetic process from aspartate
          nicotinamide-nucleotide adenylyltransferase activity
          nicotinate-nucleotide adenylyltransferase activity
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline