Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020247.4 | NP_064632.2 | 3 | 744 | ||
XM_005273201.1 | XP_005273258.1 | 3 | 640 | |||
XM_011544238.1 | XP_011542540.1 | 4 | 836 | |||
XM_011544239.2 | XP_011542541.1 | 5 | 859 | |||
XM_011544240.2 | XP_011542542.1 | 3 | 653 | |||
XM_011544241.2 | XP_011542543.1 | 3 | 615 | |||
XM_017001852.1 | XP_016857341.1 | 3 | 834 | |||
GenBank mRNA | AB073905.1 | - | 3 | 667 | ||
AK074693.1 | - | 3 | 687 | |||
AK090494.1 | - | 3 | 481 | |||
AK092784.1 | - | 3 | 467 | |||
AK293571.1 | - | 4 | 641 | |||
AK301012.1 | - | 5 | 665 | |||
BC005171.2 | - | 3 | 693 | 3604948 | ||
BC129930.1 | - | 9 | 1586 | 40010265 | ||
BX648860.1 | - | 3 | 565 |