Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APMAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020531.2 | NP_065392.1 | 8 | 1118 | ||
GenBank mRNA | AB033767.1 | - | 8 | 1057 | ||
AK025834.1 | - | 4 | 582 | |||
AK026866.1 | - | 5 | 616 | |||
AK291129.1 | - | 8 | 1079 | |||
AY359076.1 | - | 7 | 905 | |||
BC003501.1 | - | 8 | 1042 | 3530962 |