Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LYRM1 SIRNA Assays ›
Gene Symbol:
LYRM1
Gene Name:
LYR motif containing 1
Gene Aliases:
A211C6.1
Chromosome Location:
Chr.16: 20899626 - 20925006 on Build GRCh38
UniGene ID:
Hs.745012
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s32786
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LYRM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128301.2
NP_001121773.1
4
635
NM_001128302.2
NP_001121774.1
3
403
NM_001302835.1
NP_001289764.1
5
579
NM_001302836.1
NP_001289765.1
5
731
NM_020424.4
NP_065157.1
4
554
NR_126516.1
3
476
NR_126520.1
2
258
NR_126521.1
2
244
NR_126527.1
2
72
NR_126528.1
3
490
XM_005255444.1
XP_005255501.1
4
456
XM_011545900.2
XP_011544202.1
4
218
XM_011545901.2
XP_011544203.1
4
284
XM_011545902.1
XP_011544204.1
5
555
XM_011545908.2
XP_011544210.1
4
286
XM_017023475.1
XP_016878964.1
5
971
XM_017023477.1
XP_016878966.1
3
806
XM_017023478.1
XP_016878967.1
4
276
XM_017023479.1
XP_016878968.1
4
713
XM_017023480.1
XP_016878969.1
6
799
XM_017023481.1
XP_016878970.1
4
428
XM_017023482.1
XP_016878971.1
3
482
GenBank mRNA
AK308690.1
-
2
323
AK310055.1
-
3
304
AK311321.1
-
4
456
AK311415.1
-
3
478
AK311881.1
-
3
270
AV714321.1
-
3
249
BC017039.1
-
5
579
3849248
BF575136.1
-
3
292
BG541469.1
-
2
107
BI547471.1
-
3
321
BI825882.1
-
5
578
BM546065.1
-
2
208
BX096767.1
-
2
72
CD237780.1
-
3
273
CX163936.1
-
3
280
DB054440.1
-
4
491
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0