Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167882.1 | NP_001161354.1 | 3 | 3915 | ||
NM_020337.2 | NP_065070.1 | 4 | 5190 | |||
XM_017008471.1 | XP_016863960.1 | 3 | 5923 | |||
GenBank mRNA | AB033049.2 | - | 1 | 3134 | ||
AK091516.1 | - | 1 | 3207 | |||
AK127496.1 | - | 3 | 4041 | |||
AK291068.1 | - | 4 | 5190 | |||
AK295135.1 | - | 3 | 3915 | |||
BC024725.1 | - | 1 | 2214 | 4943808 | ||
BX640674.1 | - | 3 | 3707 | |||
KU178729.1 | - | 1 | 2094 | |||
KU178730.1 | - | 2 | 641 |