Search
Search

HHATL| Gene Symbol: HHATL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020707.3 | NP_065758.3 | 12 | 1682 | ||
| NR_027753.1 | 13 | 1567 | ||||
| XM_006713274.2 | XP_006713337.1 | 12 | 1636 | |||
| XM_006713275.2 | XP_006713338.1 | 11 | 1509 | |||
| XM_011533969.2 | XP_011532271.1 | 13 | 1716 | |||
| XM_011533970.2 | XP_011532272.1 | 12 | 1523 | |||
| XM_017006935.1 | XP_016862424.1 | 13 | 1542 | |||
| XM_017006936.1 | XP_016862425.1 | 13 | 1736 | |||
| XM_017006937.1 | XP_016862426.1 | 12 | 1367 | |||
| XM_017006938.1 | XP_016862427.1 | 11 | 1441 | |||
| GenBank mRNA | AB032999.1 | - | 7 | 907 | ||
| AB042554.1 | - | 12 | 1489 | |||
| AF109356.1 | - | 9 | 1156 | |||
| BC022529.1 | - | 13 | 1567 | 4799816 | ||