Search
Search

HHATL| Gene Symbol: HHATL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020707.3 | NP_065758.3 | 10 | 1429 | ||
| NR_027753.1 | 11 | 1314 | ||||
| XM_006713274.2 | XP_006713337.1 | 10 | 1383 | |||
| XM_006713275.2 | XP_006713338.1 | 9 | 1256 | |||
| XM_011533969.2 | XP_011532271.1 | 11 | 1463 | |||
| XM_011533970.2 | XP_011532272.1 | 10 | 1270 | |||
| XM_017006935.1 | XP_016862424.1 | 11 | 1289 | |||
| XM_017006936.1 | XP_016862425.1 | 11 | 1483 | |||
| XM_017006937.1 | XP_016862426.1 | 10 | 1114 | |||
| XM_017006938.1 | XP_016862427.1 | 9 | 1188 | |||
| GenBank mRNA | AB032999.1 | - | 5 | 654 | ||
| AB042554.1 | - | 10 | 1236 | |||
| AF109356.1 | - | 7 | 903 | |||
| BC022529.1 | - | 11 | 1314 | 4799816 | ||