Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MTUS1 SIRNA Assays ›
Gene Symbol:
MTUS1
Gene Name:
microtubule associated tumor suppressor 1
Gene Aliases:
ATBP, ATIP, ICIS, MP44, MTSG1
Chromosome Location:
Chr.8: 17643794 - 17801579 on Build GRCh38
UniGene ID:
Hs.7946
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s33187
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MTUS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001924.2
NP_001001924.1
15
4147
NM_001001925.2
NP_001001925.1
14
3985
NM_001001931.2
NP_001001931.1
12
1734
NM_001166393.1
NP_001159865.1
12
1379
NM_020749.4
NP_065800.1
10
1531
XM_005273577.3
XP_005273634.1
15
4055
XM_005273578.3
XP_005273635.1
15
4143
XM_005273579.3
XP_005273636.1
15
4059
XM_005273580.3
XP_005273637.1
14
3893
XM_005273581.3
XP_005273638.1
14
3981
XM_005273582.3
XP_005273639.1
14
3897
XM_011544595.2
XP_011542897.1
15
4089
XM_017013698.1
XP_016869187.1
16
4061
XM_017013699.1
XP_016869188.1
16
4283
XM_017013700.1
XP_016869189.1
15
3946
XM_017013701.1
XP_016869190.1
16
4063
XM_017013702.1
XP_016869191.1
14
3718
XM_017013704.1
XP_016869193.1
9
1053
GenBank mRNA
AB033114.1
-
14
3599
AF086371.1
-
2
375
AF121259.1
-
10
1171
AF293357.1
-
10
1444
AK000172.1
-
3
272
AK024357.1
-
9
1057
AK093875.1
-
2
1890
AK125188.1
-
12
1734
AK289750.1
-
10
1530
AK294860.1
-
12
1320
AK304105.1
-
13
1616
AK314692.1
-
10
1465
AL096842.1
-
15
2939
BC033842.1
-
7
623
BC136320.1
-
15
3898
9020307
BC142971.1
-
10
1464
BX648879.1
-
8
2037
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65b4bdbffd-d6q6k:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0