Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: PCDH19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105243.1 | NP_001098713.1 | 1 | 3790 | ||
NM_001184880.1 | NP_001171809.1 | 1 | 3790 | |||
NM_020766.2 | NP_065817.2 | 1 | 3790 | |||
XM_011530997.2 | XP_011529299.1 | 1 | 2759 | |||
GenBank mRNA | AK096591.1 | - | 1 | 743 | ||
BC136628.1 | - | 1 | 671 | 9020618 | ||
CR749278.1 | - | 1 | 671 | |||
EF676096.1 | - | 1 | 3790 |