Search
Search

IFT80| Gene Symbol: IFT80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190241.1 | NP_001177170.1 | 8 | 1252 | ||
| NM_001190242.1 | NP_001177171.1 | 6 | 886 | |||
| NM_020800.2 | NP_065851.1 | 7 | 1032 | |||
| GenBank mRNA | AB037795.1 | - | 6 | 776 | ||
| AK055904.1 | - | 7 | 1818 | |||
| AK125342.1 | - | 7 | 1032 | |||
| AK303410.1 | - | 7 | 1030 | |||
| BC101494.1 | - | 7 | 674 | 8069000 | ||
| BC113669.1 | - | 7 | 674 | 8322721 | ||