Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other USP28 SIRNA Assays ›
Gene Symbol:
USP28
Gene Name:
ubiquitin specific peptidase 28
Gene Aliases:
-
Chromosome Location:
Chr.11: 113797875 - 113875580 on Build GRCh38
UniGene ID:
Hs.503891
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s33508
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: USP28
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001301029.1
NP_001287958.1
19
2483
NM_020886.3
NP_065937.1
22
2790
XM_005271630.2
XP_005271687.1
23
2975
XM_005271631.2
XP_005271688.1
23
2972
XM_005271632.2
XP_005271689.1
24
3305
XM_005271633.4
XP_005271690.1
24
3033
XM_005271636.2
XP_005271693.1
22
2795
XM_005271637.2
XP_005271694.1
22
2789
XM_005271638.2
XP_005271695.1
21
2693
XM_005271639.2
XP_005271696.1
21
2690
XM_011542936.2
XP_011541238.1
22
2894
XM_011542938.1
XP_011541240.1
21
2768
XM_011542941.1
XP_011541243.1
16
2151
XM_011542942.1
XP_011541244.1
12
1691
XM_017018056.1
XP_016873545.1
22
2891
XM_017018057.1
XP_016873546.1
23
3116
XM_017018058.1
XP_016873547.1
21
2722
XM_017018059.1
XP_016873548.1
20
2609
XM_017018060.1
XP_016873549.1
21
2765
XM_017018061.1
XP_016873550.1
20
2582
XM_017018062.1
XP_016873551.1
20
2579
XM_017018063.1
XP_016873552.1
22
2852
XM_017018064.1
XP_016873553.1
20
2587
XM_017018065.1
XP_016873554.1
23
2831
XM_017018066.1
XP_016873555.1
24
3193
XM_017018067.1
XP_016873556.1
19
2442
XM_017018068.1
XP_016873557.1
14
1869
XM_017018069.1
XP_016873558.1
10
1409
GenBank mRNA
AB040948.1
-
12
1837
AF266283.1
-
22
2727
AK027521.1
-
3
226
AK097623.1
-
21
2324
AK304770.1
-
19
2454
CD704003.1
-
1
100
Back To Top
More Information
Panther Classification:
Molecular Function -
cysteine protease
Gene Ontology Categories:
Function(s)
Process(es)
DNA damage checkpoint
DNA repair
ubiquitin-dependent protein catabolic process
cellular response to DNA damage stimulus
Ras protein signal transduction
cell proliferation
response to ionizing radiation
protein deubiquitination
regulation of protein stability
cellular response to UV
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
thiol-dependent ubiquitin-specific protease activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0