Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBRSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142641.1 | NP_001136113.1 | 10, 11 | 1480 | ||
XM_005266171.3 | XP_005266228.1 | 11, 12 | 1660 | |||
XM_005266175.3 | XP_005266232.1 | 11, 12 | 1663 | |||
XM_005266176.3 | XP_005266233.1 | 10, 11 | 1536 | |||
XM_005266177.3 | XP_005266234.1 | 11, 12 | 1655 | |||
GenBank mRNA | AB046765.2 | - | 10, 11 | 1251 | ||
AF490258.1 | - | 3, 4 | 346 |