Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MIER1 SIRNA Assays ›
Gene Symbol:
MIER1
Gene Name:
MIER1 transcriptional regulator
Gene Aliases:
ER1, MI-ER1
Chromosome Location:
Chr.1: 66924895 - 66988619 on Build GRCh38
UniGene ID:
Hs.605432
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s33644
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MIER1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077700.2
NP_001071168.2
10, 11
1150
NM_001077701.2
NP_001071169.1
9, 10
1006
NM_001077702.2
NP_001071170.2
11, 12
1224
NM_001077703.2
NP_001071171.2
10, 11
1150
NM_001077704.2
NP_001071172.1
9, 10
1006
NM_001146110.1
NP_001139582.1
11, 12
1227
NM_001146111.1
NP_001139583.1
11, 12
1224
NM_001146112.1
NP_001139584.1
11, 12
1185
NM_001146113.1
NP_001139585.1
9, 10
1004
NM_020948.3
NP_065999.2
11, 12
1224
XM_005271076.3
XP_005271133.1
11, 12
1262
XM_011541864.2
XP_011540166.1
12, 13
1169
XM_011541865.2
XP_011540167.1
12, 13
1130
XM_017001919.1
XP_016857408.1
12, 13
1146
XM_017001920.1
XP_016857409.1
12, 13
1163
XM_017001921.1
XP_016857410.1
12, 13
1191
XM_017001922.1
XP_016857411.1
12, 13
1166
XM_017001923.1
XP_016857412.1
10, 11
986
XM_017001924.1
XP_016857413.1
11, 12
1118
XM_017001925.1
XP_016857414.1
11, 12
1223
XM_017001926.1
XP_016857415.1
12, 13
1162
XM_017001927.1
XP_016857416.1
10, 11
1035
XM_017001928.1
XP_016857417.1
10, 11
1111
XM_017001929.1
XP_016857418.1
11, 12
1092
XM_017001930.1
XP_016857419.1
10, 11
1036
XM_017001931.1
XP_016857420.1
11, 12
1070
XM_017001932.1
XP_016857421.1
9, 10
999
XM_017001933.1
XP_016857422.1
8, 9
853
XM_017001934.1
XP_016857423.1
7, 8
749
GenBank mRNA
AB046830.1
-
7, 8
775
AF515446.1
-
9, 10
1006
AF515447.1
-
10, 11
1087
AF515448.2
-
11, 12
1161
AK302061.1
-
10, 11
1150
AK307717.1
-
11, 12
1121
AL831987.2
-
11, 12
1166
AY124186.2
-
11, 12
1161
AY124187.1
-
10, 11
1087
AY124188.1
-
9, 10
1006
AY124189.2
-
11, 12
1161
AY124190.1
-
10, 11
1087
AY124191.1
-
9, 10
1006
AY124192.2
-
11, 12
1161
AY124193.1
-
10, 11
1087
AY124194.1
-
9, 10
1006
BC108726.1
-
11, 12
1105
5194296
BC125217.1
-
8, 9
802
40123797
BC125218.1
-
8, 9
802
40123803
CR627441.1
-
11, 12
1061
Back To Top
More Information
Panther Classification:
Molecular Function -
homeodomain transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
signal transduction
positive regulation of chromatin silencing
positive regulation of I-kappaB kinase/NF-kappaB signaling
DNA binding
signal transducer activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9fjjm:80/10.239.165.64:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0