Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317924.1 | NP_001304853.1 | 8 | 774 | ||
NM_025132.3 | NP_079408.3 | 9 | 890 | |||
XM_011513724.1 | XP_011512026.1 | 9 | 800 | |||
XM_011513725.2 | XP_011512027.1 | 8 | 755 | |||
XM_011513726.2 | XP_011512028.1 | 8 | 663 | |||
XM_011513727.1 | XP_011512029.1 | 7 | 583 | |||
XM_017008501.1 | XP_016863990.1 | 7 | 583 | |||
GenBank mRNA | AK026780.1 | - | 9 | 765 | ||
AK225588.1 | - | 9 | 765 | |||
AK294730.1 | - | 8 | 649 | |||
AK304161.1 | - | 9 | 763 | |||
AY029257.1 | - | 9 | 890 | |||
BC032578.1 | - | 8 | 769 |