Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANKRD36 SIRNA Assays ›
          Gene Symbol:
          ANKRD36 ANKRD36B
          Gene Name:
          ankyrin repeat domain 36
          ankyrin repeat domain 36B
          Gene Aliases:
          UNQ2430
          KIAA1641
          Chromosome Location:
          Chr.2: 97113186 - 97264521 on Build GRCh38
          Chromosome Location:
          Chr.2: 97492663 - 97590245 on Build GRCh38
          UniGene ID:
          Hs.541894
          Hs.532921
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s33696

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ANKRD36
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164315.1 NP_001157787.1 71 5236
          XM_006712514.3 XP_006712577.1 71 5468
          XM_011511130.1 XP_011509432.1 71 5468
          XM_017004009.1 XP_016859498.1 71 5468
          XM_017004010.1 XP_016859499.1 71 5465
          XM_017004011.1 XP_016859500.1 69 5366
          XM_017004012.1 XP_016859501.1 69 5366
          XM_017004013.1 XP_016859502.1 69 5366
          XM_017004014.1 XP_016859503.1 69 5366
          XM_017004015.1 XP_016859504.1 69 5366
          XM_017004016.1 XP_016859505.1 69 5366
          XM_017004017.1 XP_016859506.1 69 5366
          XM_017004018.1 XP_016859507.1 69 5366
          XM_017004019.1 XP_016859508.1 69 5366
          XM_017004020.1 XP_016859509.1 69 5366
          XM_017004021.1 XP_016859510.1 69 5366
          XM_017004022.1 XP_016859511.1 69 5366
          XM_017004023.1 XP_016859512.1 69 5366
          XM_017004024.1 XP_016859513.1 70 5330
          XM_017004025.1 XP_016859514.1 67 5264
          XM_017004026.1 XP_016859515.1 67 5264
          XM_017004027.1 XP_016859516.1 67 5264
          XM_017004028.1 XP_016859517.1 67 5264
          XM_017004029.1 XP_016859518.1 67 5264
          XM_017004030.1 XP_016859519.1 67 5264
          XM_017004031.1 XP_016859520.1 67 5264
          XM_017004032.1 XP_016859521.1 67 5264
          XM_017004033.1 XP_016859522.1 67 5264
          XM_017004034.1 XP_016859523.1 67 5264
          XM_017004035.1 XP_016859524.1 67 5264
          XM_017004036.1 XP_016859525.1 67 5264
          XM_017004037.1 XP_016859526.1 67 5264
          XM_017004038.1 XP_016859527.1 67 5264
          XM_017004039.1 XP_016859528.1 65 5162
          XM_017004040.1 XP_016859529.1 65 5162
          XM_017004041.1 XP_016859530.1 65 5162
          XM_017004042.1 XP_016859531.1 65 5162
          XM_017004043.1 XP_016859532.1 65 5162
          XM_017004044.1 XP_016859533.1 63 5060
          XM_017004045.1 XP_016859534.1 63 5060
          XM_017004046.1 XP_016859535.1 69 5223
          XM_017004047.1 XP_016859536.1 68 5155
          XM_017004048.1 XP_016859537.1 67 5082
          XM_017004049.1 XP_016859538.1 69 5257
          XM_017004050.1 XP_016859539.1 54 3661
          XM_017004051.1 XP_016859540.1 52 3875
          GenBank mRNA AB046861.1 - 13 2134
          AF543495.1 - 3 677
          AK090926.1 - 1 328
          BC146835.1 - 13 2145 8860491
          Gene Symbol: ANKRD36B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_025190.3 NP_079466.3 39 3508
          XM_005263995.4 XP_005264052.1 39 3788
          XM_006712661.1 XP_006712724.1 39 3788
          XM_017004578.1 XP_016860067.1 39 3785
          XM_017004579.1 XP_016860068.1 37 3686
          XM_017004580.1 XP_016860069.1 37 3686
          XM_017004581.1 XP_016860070.1 37 3686
          XM_017004582.1 XP_016860071.1 37 3686
          XM_017004583.1 XP_016860072.1 38 3650
          XM_017004584.1 XP_016860073.1 33 3482
          XM_017004585.1 XP_016860074.1 37 3543
          XM_017004586.1 XP_016860075.1 34 2529
          XM_017004587.1 XP_016860076.1 38 3717
          XM_017004588.1 XP_016860077.1 37 3582
          XM_017004589.1 XP_016860078.1 36 3475
          XM_017004590.1 XP_016860079.1 35 3402
          XM_017004591.1 XP_016860080.1 21 3506
          XM_017004592.1 XP_016860081.1 40 3832
          XM_017004593.1 XP_016860082.1 38 3638
          XM_017004594.1 XP_016860083.1 37 3531
          XM_017004595.1 XP_016860084.1 38 3587

          Back To Top

          More Information


          Gene Ontology Categories:

          Process(es)

          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0